Product Details
- SNP ID
-
rs3745653
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12577657 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCAGCAAACCTTGCTCCAGTCGG[C/T]CTCTGGACCCTAGTATCTTCACCGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF490
PubMed Links
Gene Details
- Gene
- ZNF490
- Gene Name
- zinc finger protein 490
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020714.2 |
4547 |
UTR 3 |
|
|
NP_065765.1 |
View Full Product Details