Product Details

SNP ID
rs3735204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:157539410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGGATGGAGGTGCGTTTTCTA[C/T]GCTGAACCCCACACAGGAAATCTGC
Phenotype
MIM: 601698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPRN2 PubMed Links

Gene Details

Gene
PTPRN2
Gene Name
protein tyrosine phosphatase, receptor type N2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308267.1 4485 UTR 3 NP_001295196.1
NM_001308268.1 4485 UTR 3 NP_001295197.1
NM_002847.4 4485 UTR 3 NP_002838.2
NM_130842.3 4485 UTR 3 NP_570857.2
NM_130843.3 4485 UTR 3 NP_570858.2
XM_011516446.1 4485 Intron XP_011514748.1
XM_011516447.2 4485 Intron XP_011514749.1
XM_011516448.2 4485 Intron XP_011514750.1
XM_011516449.2 4485 Intron XP_011514751.1
XM_017012475.1 4485 UTR 3 XP_016867964.1
XM_017012476.1 4485 Intron XP_016867965.1

View Full Product Details