Product Details

SNP ID
rs3752311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37517594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATCTCTCTTTCCCCTCAGGCAA[C/G]AGAGAGATGGATGGATCAGACTGAA
Phenotype
MIM: 613110 MIM: 603106
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BLCAP PubMed Links

Gene Details

Gene
BLCAP
Gene Name
bladder cancer associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167820.1 1896 UTR 3 NP_001161292.1
NM_001167821.1 1896 UTR 3 NP_001161293.1
NM_001167822.2 1896 UTR 3 NP_001161294.1
NM_001167823.1 1896 UTR 3 NP_001161295.1
NM_001317074.1 1896 UTR 3 NP_001304003.1
NM_001317075.1 1896 UTR 3 NP_001304004.1
NM_006698.3 1896 UTR 3 NP_006689.1
XM_017027599.1 1896 UTR 3 XP_016883088.1
Gene
NNAT
Gene Name
neuronatin
There are no transcripts associated with this gene.

View Full Product Details