Product Details
- SNP ID
-
rs3738211
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:167054487 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- GGTTCCTGCAGGGCAGCAGGGGACA[C/G]AGGGGAAGGATTTGCTCTCAGGTGG
- Phenotype
-
MIM: 602171
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPA33
PubMed Links
Gene Details
- Gene
- GPA33
- Gene Name
- glycoprotein A33
View Full Product Details