Product Details

SNP ID
rs3743768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:51137376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCCGTTGGAGAGCGCTGCTGC[A/G]TACTGATTCCAGAAGCTGGAAGGAT
Phenotype
MIM: 602218
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371253 PubMed Links

Gene Details

Gene
LOC105371253
Gene Name
uncharacterized LOC105371253
There are no transcripts associated with this gene.

Gene
SALL1
Gene Name
spalt like transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127892.1 3742 Silent Mutation TAC,TAT Y1140Y NP_001121364.1
NM_002968.2 3742 Silent Mutation TAC,TAT Y1237Y NP_002959.2
XM_006721241.3 3742 Silent Mutation TAC,TAT Y1237Y XP_006721304.1
XM_011523254.2 3742 Silent Mutation TAC,TAT Y1237Y XP_011521556.1

View Full Product Details