Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001092.4 | 3873 | UTR 3 | NP_001083.2 | ||
NM_001159746.2 | 3873 | UTR 3 | NP_001153218.1 | ||
NM_001256847.2 | 3873 | UTR 3 | NP_001243776.1 | ||
NM_001282149.1 | 3873 | UTR 3 | NP_001269078.1 | ||
NM_001322840.1 | 3873 | UTR 3 | NP_001309769.1 | ||
NM_001322841.1 | 3873 | UTR 3 | NP_001309770.1 | ||
NM_001322842.1 | 3873 | Intron | NP_001309771.1 | ||
NM_021962.4 | 3873 | UTR 3 | NP_068781.2 | ||
XM_011523810.2 | 3873 | UTR 3 | XP_011522112.1 | ||
XM_011523811.2 | 3873 | Intron | XP_011522113.2 | ||
XM_011523812.2 | 3873 | UTR 3 | XP_011522114.1 | ||
XM_011523813.2 | 3873 | UTR 3 | XP_011522115.1 | ||
XM_011523814.2 | 3873 | UTR 3 | XP_011522116.1 | ||
XM_011523815.2 | 3873 | UTR 3 | XP_011522117.1 | ||
XM_017024539.1 | 3873 | UTR 3 | XP_016880028.1 | ||
XM_017024540.1 | 3873 | UTR 3 | XP_016880029.1 | ||
XM_017024541.1 | 3873 | Intron | XP_016880030.1 | ||
XM_017024542.1 | 3873 | UTR 3 | XP_016880031.1 | ||
XM_017024543.1 | 3873 | UTR 3 | XP_016880032.1 |