Product Details

SNP ID
rs3746117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3762678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGACGTCTTCCGGAAACGTGCAC[A/T]TGCAAGCTGCCCGCAATACGTCATG
Phenotype
MIM: 604262 MIM: 611858 MIM: 610362
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
APBA3 PubMed Links

Gene Details

Gene
APBA3
Gene Name
amyloid beta precursor protein binding family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004886.3 12 Intron NP_004877.1
XM_006722950.3 12 Intron XP_006723013.1
XM_006722951.3 12 Intron XP_006723014.1
Gene
MRPL54
Gene Name
mitochondrial ribosomal protein L54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172251.2 12 UTR 5 NP_758455.1
Gene
RAX2
Gene Name
retina and anterior neural fold homeobox 2
There are no transcripts associated with this gene.

View Full Product Details