Product Details

SNP ID
rs3765897
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:11014525 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGCCAAGGAAATACCTAATTTCAGC[A/G]TTTTGAAGTAGTGTGCTGTTAGTAG
Phenotype
MIM: 605078
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TARDBP PubMed Links

Gene Details

Gene
TARDBP
Gene Name
TAR DNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007375.3 Intron NP_031401.1
XM_017000863.1 Intron XP_016856352.1
XM_017000864.1 Intron XP_016856353.1
XM_017000865.1 Intron XP_016856354.1
XM_017000866.1 Intron XP_016856355.1
XM_017000867.1 Intron XP_016856356.1
XM_017000868.1 Intron XP_016856357.1

View Full Product Details