Product Details
- SNP ID
-
rs3766245
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:46560693 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- TATCATGCTGTAACTTATCCCCTTG[C/T]CTACTTCTCTCCACTAGAGCACAGA
- Phenotype
-
MIM: 611455
MIM: 606724
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KNCN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142862589] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KNCN
- Gene Name
- kinocilin
There are no transcripts associated with this gene.
- Gene
- MKNK1
- Gene Name
- MAP kinase interacting serine/threonine kinase 1
- Gene
- MKNK1-AS1
- Gene Name
- MKNK1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details