Product Details

SNP ID
rs3788744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153181506 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTTATCCTGGGATCATTGATGTC[A/G]GGGACGGGGAGGCCTTGGTCTGAGA
Phenotype
MIM: 300016
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEA1 PubMed Links

Gene Details

Gene
MAGEA1
Gene Name
MAGE family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004988.4 Intron NP_004979.3

View Full Product Details