Product Details

SNP ID
rs3801271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:106256223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAAACACAACAGAAAAAGAAGA[C/T]AGGCGCTCTGTATTTTGTCCAACAC
Phenotype
MIM: 608764
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAMPT PubMed Links

Gene Details

Gene
NAMPT
Gene Name
nicotinamide phosphoribosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005746.2 Intron NP_005737.1
XM_005250100.2 Intron XP_005250157.1

View Full Product Details