Product Details

SNP ID
rs3806818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6640604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGACGTGCGCGCGCCCGCAGCTC[A/G]CCCCCCTGACCGTAAAGAGGCCCGG
Phenotype
MIM: 616905
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRFAP1 PubMed Links

Gene Details

Gene
MRFAP1
Gene Name
Morf4 family associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272053.1 514 UTR 5 NP_001258982.1
NM_001272054.1 514 UTR 5 NP_001258983.1
NM_033296.2 514 UTR 5 NP_150638.1

View Full Product Details