Product Details
- SNP ID
-
rs3810472
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63406707 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCCTCGCCGGTGGCCGAGCGTGG[C/T]GGGGGCCCGCACGGGGTACAGAGGT
- Phenotype
-
MIM: 602235
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KCNQ2
PubMed Links
Gene Details
- Gene
- KCNQ2
- Gene Name
- potassium voltage-gated channel subfamily Q member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004518.5 |
2176 |
Silent Mutation |
CCA,CCG |
P824P |
NP_004509.2 |
NM_172106.2 |
2176 |
Silent Mutation |
CCA,CCG |
P834P |
NP_742104.1 |
NM_172107.3 |
2176 |
Silent Mutation |
CCA,CCG |
P852P |
NP_742105.1 |
NM_172108.4 |
2176 |
Silent Mutation |
CCA,CCG |
P821P |
NP_742106.1 |
NM_172109.2 |
2176 |
Intron |
|
|
NP_742107.1 |
XM_011528810.1 |
2176 |
Silent Mutation |
CCA,CCG |
P870P |
XP_011527112.1 |
XM_011528811.1 |
2176 |
Silent Mutation |
CCA,CCG |
P860P |
XP_011527113.1 |
XM_017027841.1 |
2176 |
Silent Mutation |
CCA,CCG |
P869P |
XP_016883330.1 |
XM_017027842.1 |
2176 |
Silent Mutation |
CCA,CCG |
P848P |
XP_016883331.1 |
XM_017027843.1 |
2176 |
Silent Mutation |
CCA,CCG |
P847P |
XP_016883332.1 |
XM_017027844.1 |
2176 |
Silent Mutation |
CCA,CCG |
P833P |
XP_016883333.1 |
XM_017027845.1 |
2176 |
Silent Mutation |
CCA,CCG |
P524P |
XP_016883334.1 |
- Gene
- LOC100130152
- Gene Name
- uncharacterized LOC100130152
There are no transcripts associated with this gene.
View Full Product Details