Product Details

SNP ID
rs3812944
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74997624 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAGAGGAGCTGATAGTGCTGCCA[A/G]TTTAGGGGCACTGGAAGCAGAGGGG
Phenotype
MIM: 613766
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SCAMP5 PubMed Links

Gene Details

Gene
SCAMP5
Gene Name
secretory carrier membrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178111.1 Intron NP_001171582.1
NM_001178112.1 Intron NP_001171583.1
NM_138967.3 Intron NP_620417.1
XM_006720420.1 Intron XP_006720483.1
XM_006720421.1 Intron XP_006720484.1
XM_006720422.3 Intron XP_006720485.1
XM_017021989.1 Intron XP_016877478.1

View Full Product Details