Product Details

SNP ID
rs3733840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149348359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCCTCCTGATGAGCTTGGGCC[C/T]CCTCTTGCTGAACGAGCCTTAAGGG
Phenotype
MIM: 614410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AFAP1L1 PubMed Links

Gene Details

Gene
AFAP1L1
Gene Name
actin filament associated protein 1 like 1
There are no transcripts associated with this gene.

Gene
GRPEL2
Gene Name
GrpE like 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152407.3 291 Silent Mutation CCC,CCT P55P NP_689620.2
Gene
GRPEL2-AS1
Gene Name
GRPEL2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PCYOX1L
Gene Name
prenylcysteine oxidase 1 like
There are no transcripts associated with this gene.

View Full Product Details