Product Details

SNP ID
rs3803024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48997337 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGTAAAAGGCGACTGCTCGACAG[C/T]CGCTTTTGACAAGGGGAAGGAAAGA
Phenotype
MIM: 610588 MIM: 602742
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDN PubMed Links

Gene Details

Gene
DDN
Gene Name
dendrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015086.1 2162 Silent Mutation CGA,CGG R513R NP_055901.2
XM_011538055.2 2162 Silent Mutation CGA,CGG R459R XP_011536357.1
Gene
PRKAG1
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 1
There are no transcripts associated with this gene.

View Full Product Details