Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001099403.1 | 38 | Intron | NP_001092873.1 | ||
NM_020226.3 | 38 | Intron | NP_064611.3 | ||
XM_005263144.2 | 38 | Intron | XP_005263201.1 | ||
XM_005263145.2 | 38 | Intron | XP_005263202.1 | ||
XM_005263146.4 | 38 | Intron | XP_005263203.1 | ||
XM_011532133.2 | 38 | UTR 5 | XP_011530435.1 | ||
XM_011532135.2 | 38 | UTR 5 | XP_011530437.1 | ||
XM_011532140.2 | 38 | Intron | XP_011530442.1 | ||
XM_011532141.2 | 38 | Intron | XP_011530443.1 | ||
XM_017008468.1 | 38 | Intron | XP_016863957.1 | ||
XM_017008469.1 | 38 | Intron | XP_016863958.1 | ||
XM_017008470.1 | 38 | Intron | XP_016863959.1 |