Product Details
- SNP ID
-
rs3825763
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:105176302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGTCACCCGGCCTGCCTCACAC[A/G]GCTGGGGGCCGCAGGCTGGGGGCAT
- Phenotype
-
MIM: 602570
MIM: 609219
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
JAG2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142840048] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- JAG2
- Gene Name
- jagged 2
There are no transcripts associated with this gene.
- Gene
- NUDT14
- Gene Name
- nudix hydrolase 14
View Full Product Details