Product Details

SNP ID
rs3826615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63890856 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATGAGTCAAAGTCACTTTAGAAG[C/T]AGGAAAGCAGAAAGAAGATTGACCG
Phenotype
MIM: 173390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINB2 PubMed Links

Gene Details

Gene
SERPINB2
Gene Name
serpin family B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143818.1 Intron NP_001137290.1
NM_002575.2 Intron NP_002566.1

View Full Product Details