Product Details

SNP ID
rs3761567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:110674971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTGGAATTCTTGACTAAAATA[C/T]CTGCTTGATGAATAAGACAAGGATA
Phenotype
MIM: 300350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRDL1 PubMed Links

Gene Details

Gene
CHRDL1
Gene Name
chordin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143981.1 2788 UTR 3 NP_001137453.1
NM_001143982.1 2788 UTR 3 NP_001137454.1
NM_001143983.2 2788 UTR 3 NP_001137455.2
NM_145234.3 2788 UTR 3 NP_660277.2
XM_005262221.1 2788 UTR 3 XP_005262278.1
XM_005262222.3 2788 UTR 3 XP_005262279.1
XM_005262223.1 2788 UTR 3 XP_005262280.1
XM_005262224.1 2788 UTR 3 XP_005262281.1
XM_017029959.1 2788 UTR 3 XP_016885448.1

View Full Product Details