Product Details

SNP ID
rs3770959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:36362584 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGGGAGGTGTCCCTTTATCAGCA[C/T]CTCTCAGCTTCACCCACCCTGCTGC
Phenotype
MIM: 606189
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRIM1 PubMed Links

Gene Details

Gene
CRIM1
Gene Name
cysteine rich transmembrane BMP regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016441.2 Intron NP_057525.1
XM_005264357.4 Intron XP_005264414.1
XM_011532898.2 Intron XP_011531200.1
XM_011532899.2 Intron XP_011531201.1
XM_011532901.2 Intron XP_011531203.1
XM_017004258.1 Intron XP_016859747.1
XM_017004259.1 Intron XP_016859748.1
XM_017004260.1 Intron XP_016859749.1
XM_017004261.1 Intron XP_016859750.1
XM_017004262.1 Intron XP_016859751.1
Gene
LOC100288911
Gene Name
uncharacterized LOC100288911
There are no transcripts associated with this gene.

View Full Product Details