Product Details

SNP ID
rs3785433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75670953 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTTCTGTGCATTGGGAGTGTTCT[G/T]GGTAGTGGAGGTGACATGGATTTAG
Phenotype
MIM: 603781 MIM: 610218
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RECQL5 PubMed Links

Gene Details

Gene
RECQL5
Gene Name
RecQ like helicase 5
There are no transcripts associated with this gene.

Gene
SAP30BP
Gene Name
SAP30 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301839.1 Intron NP_001288768.1
NM_001301855.1 Intron NP_001288784.1
NM_013260.7 Intron NP_037392.1
XM_011524693.2 Intron XP_011522995.1
XM_011524694.2 Intron XP_011522996.1
XM_011524695.2 Intron XP_011522997.1
XM_017024545.1 Intron XP_016880034.1
XM_017024546.1 Intron XP_016880035.1
XM_017024547.1 Intron XP_016880036.1
XM_017024548.1 Intron XP_016880037.1
XM_017024549.1 Intron XP_016880038.1
XM_017024550.1 Intron XP_016880039.1

View Full Product Details