Product Details

SNP ID
rs3790127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10530585 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGCTTATCTCCTGGGATGGTTCC[C/T]GGGTGGTCTGCATTGCCACACAGCA
Phenotype
MIM: 602334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMP2 PubMed Links

Gene Details

Gene
EMP2
Gene Name
epithelial membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001424.5 3046 UTR 3 NP_001415.1
XM_006720864.3 3046 UTR 3 XP_006720927.1

View Full Product Details