Product Details

SNP ID
rs3208404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30730844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAATCCATAGGAGTCCAGGTGG[C/T]GAAGGCTTCAGCACTCCATCTTGGG
Phenotype
MIM: 606998 MIM: 191130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLOT1 PubMed Links

Gene Details

Gene
FLOT1
Gene Name
flotillin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318875.1 Intron NP_001305804.1
NM_005803.3 Intron NP_005794.1
XM_005248780.3 Intron XP_005248837.1
XM_006714947.3 Intron XP_006715010.1
XM_017010157.1 Intron XP_016865646.1
XM_017010158.1 Intron XP_016865647.1
Gene
TUBB
Gene Name
tubulin beta class I
There are no transcripts associated with this gene.

View Full Product Details