Product Details

SNP ID
rs3735281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141658595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTAATTGTCTTGCCCATGTTAG[A/T]CCTGTCATCACACTAAGGCAAGTCC
Phenotype
MIM: 610345
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AGK PubMed Links

Gene Details

Gene
AGK
Gene Name
acylglycerol kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018238.3 5429 Intron NP_060708.1
Gene
KIAA1147
Gene Name
KIAA1147
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080392.1 5429 UTR 3 NP_001073861.1

View Full Product Details