Product Details
- SNP ID
-
rs3739736
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:104095627 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCACTGAACTTTGGAGACGTCCC[A/G]ATATTCTCTTCATTTGTGTTTTTAT
- Phenotype
-
MIM: 605576
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SMC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112658186] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SMC2
- Gene Name
- structural maintenance of chromosomes 2
- Gene
- SMC2-AS1
- Gene Name
- SMC2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details