Product Details

SNP ID
rs3745623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46634757 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTAGGTTAGGATACGTCTGGGTC[C/G]CGAGGTGGGGGCGAGAGGGCGGGCC
Phenotype
MIM: 600022
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GNG8 PubMed Links

Gene Details

Gene
GNG8
Gene Name
G protein subunit gamma 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033258.1 Intron NP_150283.1
XM_011527524.2 Intron XP_011525826.1
XM_017027503.1 Intron XP_016882992.1
XM_017027504.1 Intron XP_016882993.1
XM_017027505.1 Intron XP_016882994.1
Gene
PTGIR
Gene Name
prostaglandin I2 (prostacyclin) receptor (IP)
There are no transcripts associated with this gene.

View Full Product Details