Product Details
- SNP ID
-
rs3749631
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:3596404 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGTGAGTGCGCCCGGCCTCCCCC[G/T]CTTCTCCTCTGTCTCACCCGCGCCA
- Phenotype
-
MIM: 606197
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IRX1
PubMed Links
Gene Details
- Gene
- IRX1
- Gene Name
- iroquois homeobox 1
View Full Product Details