Product Details

SNP ID
rs3767870
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:212440484 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GTCTGCTGGCTTGGAAATCCAGGCC[A/G]TTTCAAATGACCCGTGTTAGATGAG
Phenotype
MIM: 611874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NENF PubMed Links

Gene Details

Gene
NENF
Gene Name
neudesin neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013349.4 Intron NP_037481.1

View Full Product Details