Product Details

SNP ID
rs3771534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69837303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGTCTGCCTCCCATGCTGCCC[C/T]CCTTTCTCCACTATCTCTCCCGTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GMCL1 PubMed Links

Gene Details

Gene
GMCL1
Gene Name
germ cell-less, spermatogenesis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178439.4 Intron NP_848526.1
XM_011533033.1 Intron XP_011531335.1
XM_011533034.2 Intron XP_011531336.1
XM_017004704.1 Intron XP_016860193.1
XM_017004705.1 Intron XP_016860194.1

View Full Product Details