Product Details

SNP ID
rs3778198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83554262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGTCTCCAAACTCATTTATTTT[C/G]CTATTCAGTCACAAATATTGCAGCT
Phenotype
MIM: 607923
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SNAP91 PubMed Links

Gene Details

Gene
SNAP91
Gene Name
synaptosome associated protein 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242792.1 2795 UTR 3 NP_001229721.1
NM_001242793.1 2795 UTR 3 NP_001229722.1
NM_001242794.1 2795 UTR 3 NP_001229723.1
NM_001256717.1 2795 UTR 3 NP_001243646.1
NM_001256718.1 2795 UTR 3 NP_001243647.1
NM_014841.2 2795 UTR 3 NP_055656.1
XM_005248770.4 2795 UTR 3 XP_005248827.1
XM_006715615.1 2795 UTR 3 XP_006715678.1
XM_011536265.1 2795 UTR 3 XP_011534567.1
XM_011536266.1 2795 UTR 3 XP_011534568.1
XM_011536269.1 2795 UTR 3 XP_011534571.1
XM_011536271.1 2795 UTR 3 XP_011534573.1
XM_011536273.2 2795 UTR 3 XP_011534575.1
XM_011536275.2 2795 UTR 3 XP_011534577.1
XM_011536276.2 2795 UTR 3 XP_011534578.1
XM_011536277.1 2795 UTR 3 XP_011534579.1
XM_017011554.1 2795 UTR 3 XP_016867043.1
XM_017011555.1 2795 UTR 3 XP_016867044.1
XM_017011556.1 2795 UTR 3 XP_016867045.1
XM_017011557.1 2795 UTR 3 XP_016867046.1
XM_017011558.1 2795 UTR 3 XP_016867047.1
XM_017011559.1 2795 UTR 3 XP_016867048.1
XM_017011560.1 2795 UTR 3 XP_016867049.1
XM_017011561.1 2795 UTR 3 XP_016867050.1
XM_017011562.1 2795 UTR 3 XP_016867051.1
XM_017011563.1 2795 UTR 3 XP_016867052.1
XM_017011564.1 2795 UTR 3 XP_016867053.1
XM_017011565.1 2795 UTR 3 XP_016867054.1
XM_017011566.1 2795 UTR 3 XP_016867055.1
XM_017011567.1 2795 UTR 3 XP_016867056.1
XM_017011568.1 2795 UTR 3 XP_016867057.1
XM_017011569.1 2795 UTR 3 XP_016867058.1
XM_017011570.1 2795 UTR 3 XP_016867059.1
XM_017011571.1 2795 UTR 3 XP_016867060.1
XM_017011572.1 2795 UTR 3 XP_016867061.1
XM_017011573.1 2795 UTR 3 XP_016867062.1
XM_017011574.1 2795 UTR 3 XP_016867063.1
XM_017011575.1 2795 UTR 3 XP_016867064.1
XM_017011576.1 2795 UTR 3 XP_016867065.1
XM_017011577.1 2795 UTR 3 XP_016867066.1
XM_017011578.1 2795 UTR 3 XP_016867067.1
XM_017011579.1 2795 UTR 3 XP_016867068.1
XM_017011580.1 2795 UTR 3 XP_016867069.1
XM_017011581.1 2795 UTR 3 XP_016867070.1
XM_017011582.1 2795 UTR 3 XP_016867071.1
XM_017011583.1 2795 UTR 3 XP_016867072.1
XM_017011584.1 2795 UTR 3 XP_016867073.1
XM_017011585.1 2795 UTR 3 XP_016867074.1
XM_017011586.1 2795 UTR 3 XP_016867075.1
XM_017011587.1 2795 UTR 3 XP_016867076.1
XM_017011588.1 2795 UTR 3 XP_016867077.1
XM_017011589.1 2795 UTR 3 XP_016867078.1
XM_017011590.1 2795 UTR 3 XP_016867079.1

View Full Product Details