Product Details

SNP ID
rs3795575
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:204155961 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GAGAGCCTTCTTGAGTATGGAAGAC[A/G]TCTCAGCAGACAAGGAGTCCTGCGC
Phenotype
MIM: 609859 MIM: 179820
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ETNK2 PubMed Links

Gene Details

Gene
ETNK2
Gene Name
ethanolamine kinase 2
There are no transcripts associated with this gene.

Gene
REN
Gene Name
renin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000537.3 Intron NP_000528.1

View Full Product Details