Product Details

SNP ID
rs3803538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90348539 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACTTGTCCACCTCCTGACCGACC[C/T]TTCTATGAAACTGCACAAAAATTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABARAPL3 PubMed Links

Gene Details

Gene
GABARAPL3
Gene Name
GABA type A receptor associated protein like 3 pseudogene
There are no transcripts associated with this gene.

Gene
ZNF774
Gene Name
zinc finger protein 774
There are no transcripts associated with this gene.

View Full Product Details