Product Details

SNP ID
rs3803866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44193609 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGCACCCAACAAAAGGACAGCT[C/T]CTGCTGCCAAGGAGGCCCATGGGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASB16-AS1 PubMed Links

Gene Details

Gene
ASB16-AS1
Gene Name
ASB16 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ATXN7L3
Gene Name
ataxin 7 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098833.1 2039 Intron NP_001092303.1
NM_020218.1 2039 Intron NP_064603.1
XM_011525038.2 2039 Intron XP_011523340.1
XM_011525044.2 2039 Intron XP_011523346.1
XM_011525046.2 2039 Intron XP_011523348.1
XM_017024880.1 2039 UTR 3 XP_016880369.1
XM_017024881.1 2039 UTR 3 XP_016880370.1
XM_017024882.1 2039 Intron XP_016880371.1
XM_017024883.1 2039 UTR 3 XP_016880372.1
XM_017024884.1 2039 UTR 3 XP_016880373.1
XM_017024885.1 2039 UTR 3 XP_016880374.1
XM_017024886.1 2039 UTR 3 XP_016880375.1
Gene
LOC101926967
Gene Name
uncharacterized LOC101926967
There are no transcripts associated with this gene.

Gene
TMUB2
Gene Name
transmembrane and ubiquitin like domain containing 2
There are no transcripts associated with this gene.

View Full Product Details