Product Details

SNP ID
rs3810177
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49031767 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGCCCGAGGGCACGCGCAACCGG[C/T]CGGAGCCACAATAGCTCGGGGTGTC
Phenotype
MIM: 608823 MIM: 608824 MIM: 118860
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CGB1 PubMed Links
Additional Information
For this assay, SNP(s) [rs116454573,rs576717480] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CGB1
Gene Name
chorionic gonadotropin beta subunit 1
There are no transcripts associated with this gene.

Gene
CGB2
Gene Name
chorionic gonadotropin beta subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319065.1 Intron NP_001305994.1
NM_033378.1 Intron NP_203696.2
XM_005258479.3 Intron XP_005258536.1
Gene
CGB3
Gene Name
chorionic gonadotropin beta subunit 3
There are no transcripts associated with this gene.

Gene
SNAR-G1
Gene Name
small ILF3/NF90-associated RNA G1
There are no transcripts associated with this gene.

Gene
SNAR-G2
Gene Name
small ILF3/NF90-associated RNA G2
There are no transcripts associated with this gene.

View Full Product Details