Product Details

SNP ID
rs4077409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88461849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACTGCTAAGATCATCGTTGGCA[C/T]TGGCCAACCCATTTTACAGGAGAGA
Phenotype
MIM: 601950
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR5189 PubMed Links

Gene Details

Gene
MIR5189
Gene Name
microRNA 5189
There are no transcripts associated with this gene.

Gene
ZFPM1
Gene Name
zinc finger protein, FOG family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153813.2 Intron NP_722520.2
XM_011522912.2 Intron XP_011521214.1
XM_011522914.2 Intron XP_011521216.1
XM_011522917.2 Intron XP_011521219.1
XM_017022982.1 Intron XP_016878471.1

View Full Product Details