Product Details
- SNP ID
-
hCV27529573
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:78780407 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCTCCCTGAAATCCTACATCCAA[C/G]GGCCCCACACCCCAGTGGCCTTGGG
- Phenotype
-
MIM: 611542
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARSB
PubMed Links
Gene Details
- Gene
- ARSB
- Gene Name
- arylsulfatase B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198709.2 |
|
Intron |
|
|
NP_942002.1 |
View Full Product Details