Product Details

SNP ID
rs121908210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6614608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGTGTTCCCCAGCCTGTTACAG[A/G]ATCCCAGCCAGGACCAGAGCAGAAA
Phenotype
MIM: 603057 MIM: 602366 MIM: 600475 MIM: 607998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCHS1 PubMed Links

Gene Details

Gene
DCHS1
Gene Name
dachsous cadherin-related 1
There are no transcripts associated with this gene.

Gene
ILK
Gene Name
integrin linked kinase
There are no transcripts associated with this gene.

Gene
TAF10
Gene Name
TATA-box binding protein associated factor 10
There are no transcripts associated with this gene.

Gene
TPP1
Gene Name
tripeptidyl peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000391.3 1691 Missense Mutation CCT,TCT P544S NP_000382.3

View Full Product Details