Product Details
- SNP ID
-
rs74315336
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:171636173 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAAGGCATTGGCGACTGACTGCT[T/C]ACGGATGTTTGTCTCCCAGGTTTGT
- Phenotype
-
MIM: 601652
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYOC
PubMed Links
Gene Details
- Gene
- MYOC
- Gene Name
- myocilin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000261.1 |
1289 |
Missense Mutation |
AAG,GAG |
K423E |
NP_000252.1 |
View Full Product Details