Product Details

SNP ID
rs72785469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:11183415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAATACGATCACCTTGAATAA[C/T]GACTGCTTTCATAGAAGGCAGTTAG
Phenotype
MIM: 604002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PQLC3 PubMed Links

Gene Details

Gene
PQLC3
Gene Name
PQ loop repeat containing 3
There are no transcripts associated with this gene.

Gene
ROCK2
Gene Name
Rho associated coiled-coil containing protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321643.1 4241 UTR 3 NP_001308572.1
NM_004850.4 4241 UTR 3 NP_004841.2
XM_005246190.3 4241 UTR 3 XP_005246247.1
XM_011510417.2 4241 UTR 3 XP_011508719.1
XM_017005378.1 4241 Intron XP_016860867.1
XM_017005379.1 4241 UTR 3 XP_016860868.1

View Full Product Details