Product Details

SNP ID
rs763761325
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:111227681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGACAGGCTGTCGAAACCTCAGT[A/G]GATAAAAGACCTAGAGAATGTGTAT
Phenotype
MIM: 601526
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHI3L2 PubMed Links

Gene Details

Gene
CHI3L2
Gene Name
chitinase 3 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025197.1 23 UTR 5 NP_001020368.1
NM_001025199.1 23 Intron NP_001020370.1
NM_004000.2 23 UTR 5 NP_003991.2

View Full Product Details