Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278508.2 | 3149 | UTR 3 | NP_001265437.2 | ||
NM_001278509.2 | 3149 | UTR 3 | NP_001265438.2 | ||
NM_001288759.2 | 3149 | UTR 3 | NP_001275688.2 | ||
NM_001288760.2 | 3149 | UTR 3 | NP_001275689.1 | ||
NM_001288761.2 | 3149 | UTR 3 | NP_001275690.1 | ||
NM_001288762.2 | 3149 | UTR 3 | NP_001275691.1 | ||
NM_001291633.1 | 3149 | UTR 3 | NP_001278562.1 | ||
NM_013256.5 | 3149 | UTR 3 | NP_037388.3 | ||
XM_011527280.2 | 3149 | UTR 3 | XP_011525582.1 | ||
XM_017027250.1 | 3149 | UTR 3 | XP_016882739.1 | ||
XM_017027251.1 | 3149 | UTR 3 | XP_016882740.1 | ||
XM_017027252.1 | 3149 | UTR 3 | XP_016882741.1 | ||
XM_017027253.1 | 3149 | UTR 3 | XP_016882742.1 | ||
XM_017027254.1 | 3149 | UTR 3 | XP_016882743.1 | ||
XM_017027255.1 | 3149 | UTR 3 | XP_016882744.1 | ||
XM_017027256.1 | 3149 | UTR 3 | XP_016882745.1 |