Product Details

SNP ID
rs7124442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:27655494 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGAAGCTGCATAAAGTTGACATA[C/T]AGCAGATATTCCAAGCATTCCTTAC
Phenotype
MIM: 113505 MIM: 611468
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BDNF PubMed Links

Gene Details

Gene
BDNF
Gene Name
brain derived neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143805.1 4155 UTR 3 NP_001137277.1
NM_001143806.1 4155 UTR 3 NP_001137278.1
NM_001143807.1 4155 UTR 3 NP_001137279.1
NM_001143808.1 4155 UTR 3 NP_001137280.1
NM_001143809.1 4155 UTR 3 NP_001137281.1
NM_001143810.1 4155 UTR 3 NP_001137282.1
NM_001143811.1 4155 UTR 3 NP_001137283.1
NM_001143812.1 4155 UTR 3 NP_001137284.1
NM_001143813.1 4155 UTR 3 NP_001137285.1
NM_001143814.1 4155 UTR 3 NP_001137286.1
NM_001143816.1 4155 UTR 3 NP_001137288.1
NM_001709.4 4155 UTR 3 NP_001700.2
NM_170731.4 4155 UTR 3 NP_733927.1
NM_170732.4 4155 UTR 3 NP_733928.1
NM_170733.3 4155 UTR 3 NP_733929.1
NM_170734.3 4155 UTR 3 NP_733930.1
NM_170735.5 4155 UTR 3 NP_733931.1
XM_011520280.2 4155 UTR 3 XP_011518582.1
Gene
BDNF-AS
Gene Name
BDNF antisense RNA
There are no transcripts associated with this gene.

View Full Product Details