Product Details

SNP ID
rs76222688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23569216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTCTTCATTTTCTTCCTCCTC[G/T]CCATTCAGCTTCCAGGAGTCTCCTT
Phenotype
MIM: 603534 MIM: 611612
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AP1G2 PubMed Links

Gene Details

Gene
AP1G2
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282474.1 3223 Intron NP_001269403.1
NM_001282475.1 3223 Intron NP_001269404.1
NM_003917.4 3223 Intron NP_003908.1
XM_005268167.3 3223 Intron XP_005268224.1
XM_005268168.4 3223 Intron XP_005268225.1
XM_005268169.3 3223 Intron XP_005268226.1
XM_005268170.3 3223 Intron XP_005268227.1
XM_005268172.3 3223 Intron XP_005268229.1
XM_005268173.3 3223 Intron XP_005268230.1
XM_005268174.3 3223 Intron XP_005268231.1
XM_005268175.3 3223 Intron XP_005268232.1
XM_005268177.3 3223 Intron XP_005268234.1
XM_005268178.3 3223 Intron XP_005268235.1
XM_005268179.3 3223 Intron XP_005268236.1
XM_005268180.4 3223 Intron XP_005268237.1
XM_005268182.3 3223 Intron XP_005268239.1
XM_006720301.3 3223 Intron XP_006720364.1
XM_011537283.2 3223 Intron XP_011535585.1
XM_011537286.2 3223 Intron XP_011535588.1
XM_017021740.1 3223 Intron XP_016877229.1
XM_017021741.1 3223 Intron XP_016877230.1
XM_017021742.1 3223 Intron XP_016877231.1
XM_017021743.1 3223 Intron XP_016877232.1
XM_017021744.1 3223 Intron XP_016877233.1
XM_017021745.1 3223 Intron XP_016877234.1
XM_017021746.1 3223 Intron XP_016877235.1
XM_017021747.1 3223 Intron XP_016877236.1
XM_017021748.1 3223 Intron XP_016877237.1
Gene
JPH4
Gene Name
junctophilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146028.1 3223 UTR 3 NP_001139500.1
NM_032452.2 3223 UTR 3 NP_115828.2
Gene
LOC102724814
Gene Name
uncharacterized LOC102724814
There are no transcripts associated with this gene.

Gene
THTPA
Gene Name
thiamine triphosphatase
There are no transcripts associated with this gene.

View Full Product Details