Product Details

SNP ID
rs116082839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74953856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTTGTTTTGGCTATTAAAGTTG[A/T]TATGTGGTGTTAAACCATGCTTTCC
Phenotype
MIM: 609788
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SPCS2 PubMed Links

Gene Details

Gene
SPCS2
Gene Name
signal peptidase complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014752.2 Intron NP_055567.2
Gene
XRRA1
Gene Name
X-ray radiation resistance associated 1
There are no transcripts associated with this gene.

View Full Product Details