Product Details

SNP ID
rs548537862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:152835720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCGATGCTTGCTGCGCCCCTGG[C/G]CGCCGCTGCCCTCTCGCCGCCTCCT
Phenotype
MIM: 601167
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
P2RY1 PubMed Links

Gene Details

Gene
P2RY1
Gene Name
purinergic receptor P2Y1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002563.4 1028 UTR 5 NP_002554.1

View Full Product Details