Product Details

SNP ID
rs80272855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6658085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGCACTGGACCGGCTCAAGGT[C/G]GGTGCCTGTGGCTGTCATGTTGGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALOX15P1 PubMed Links

Gene Details

Gene
ALOX15P1
Gene Name
arachidonate 15-lipoxygenase pseudogene 1
There are no transcripts associated with this gene.

Gene
C17orf100
Gene Name
chromosome 17 open reading frame 100
There are no transcripts associated with this gene.

Gene
MED31
Gene Name
mediator complex subunit 31
There are no transcripts associated with this gene.

Gene
MIR4520-1
Gene Name
microRNA 4520-1
There are no transcripts associated with this gene.

Gene
MIR4520-2
Gene Name
microRNA 4520-2
There are no transcripts associated with this gene.

View Full Product Details