Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001099695.1 | 256 | Missense Mutation | AGG,GGG | R34G | NP_001093165.1 |
NM_001099696.2 | 256 | Intron | NP_001093166.1 | ||
NM_013400.3 | 256 | UTR 5 | NP_037532.2 | ||
NM_014374.3 | 256 | Intron | NP_055189.2 | ||
XM_005249985.1 | 256 | UTR 5 | XP_005250042.1 | ||
XM_006715947.3 | 256 | Missense Mutation | AGG,GGG | R34G | XP_006716010.1 |
XM_006715948.3 | 256 | Missense Mutation | AGG,GGG | R34G | XP_006716011.1 |
XM_006715949.3 | 256 | Missense Mutation | AGG,GGG | R34G | XP_006716012.1 |
XM_006715950.3 | 256 | Missense Mutation | AGG,GGG | R34G | XP_006716013.1 |
XM_006715951.2 | 256 | Missense Mutation | AGG,GGG | R34G | XP_006716014.1 |
XM_006715952.2 | 256 | UTR 5 | XP_006716015.1 | ||
XM_006715953.2 | 256 | Intron | XP_006716016.1 | ||
XM_011516112.1 | 256 | UTR 5 | XP_011514414.1 | ||
XM_017012081.1 | 256 | UTR 5 | XP_016867570.1 | ||
XM_017012082.1 | 256 | UTR 5 | XP_016867571.1 |