Product Details

SNP ID
rs143646296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16555132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGGGCCGGCTTCCTTCCCTGC[C/T]CAGCCTCTCAGACTCTCACCTTGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35E1 PubMed Links

Gene Details

Gene
SLC35E1
Gene Name
solute carrier family 35 member E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024881.4 Intron NP_079157.3

View Full Product Details