Product Details

SNP ID
rs72864268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42879557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCACAGAGCGAAAATGTTTCGC[A/G]GAGTTTGCCTGTCTTCCTCCCCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL7L1 PubMed Links

Gene Details

Gene
RPL7L1
Gene Name
ribosomal protein L7 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198486.3 395 Intron NP_940888.3
XM_017010780.1 395 UTR 5 XP_016866269.1

View Full Product Details